
4-(4-Nitrobenzyl)pyridine

Synonym(s):
4-(4-Nitrobenzyl)pyridine
Linear Formula:
4-[4-(NO2)C6H4CH2]C5H4N
CAS Number:
Molecular Weight:
214.22
Products
Brand
Formula Weight
Physical Form
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing