4-[3-(Methylamino)-1-(thiophen-2-yl)propyl]naphthalen-1-ol hydrochloride
Synonym(s):
4-[3-(Methylamino)-1-(2-thienyl)propyl]-1-naphthol hydrochloride
Empirical Formula (Hill Notation):
C18H19NOS· HCl
CAS Number:
Molecular Weight:
333.88
Products
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1