Skip to Content
Merck
3-Propylidenephthalide mixture of isomers, analytical standard

3-Propylidenephthalide

3-Propylidenephthalide mixture of isomers, analytical standard
Synonym(s):
3-Propylidene-1(3H)-isobenzofuranone
Empirical Formula (Hill Notation):
C11H10O2
CAS Number:
Molecular Weight:
174.20
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Purity
Agency
Format
Quality Segment
Application
Organoleptic Class
Special Grade