Skip to Content
Merck
3-(Methylthio)-1-propanol 98%

3-(Methylthio)-1-propanol

3-(Methylthio)-1-propanol 98%
Synonym(s):
Methionol
Linear Formula:
CH3S(CH2)3OH
CAS Number:
Molecular Weight:
106.19
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing