Skip to Content
Merck
2-Phenylglycine 95%

2-Phenylglycine

2-Phenylglycine 95%
Synonym(s):
DL-α-Phenylglycine, (±)-α-Aminophenylacetic acid
Linear Formula:
C6H5CH(NH2)COOH
CAS Number:
Molecular Weight:
151.16
EC Number:

Products

Product Category
Brand
Color
Formula Weight
Melting Point (°C)
Physical Form
Purity
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Amino Acid Type
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing