Skip to Content
Merck
2-Methoxy-3(5 or 6)-isopropylpyrazine ≥98%, FG

2-Methoxy-3(5 or 6)-isopropylpyrazine

2-Methoxy-3(5 or 6)-isopropylpyrazine ≥98%, FG
Synonym(s):
3,5 Or 6-Isopropyl-2-methoxypyrazine
Empirical Formula (Hill Notation):
C8H12N2O
CAS Number:
Molecular Weight:
152.19
EC Number:

Products

Product Category
Brand
Biological Source
Formula Weight
Purity
Agency
Quality Segment
Application
Organoleptic Class
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing