Skip to Content
Merck
2,4,6-Trichlorophenol certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

2,4,6-Trichlorophenol

2,4,6-Trichlorophenol certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Synonym(s):
2,4,6-Trichlorophenol
Linear Formula:
Cl3C6H2OH
CAS Number:
Molecular Weight:
197.45
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique