![2,2′-Methylenebis(6-tert-butyl-4-methylphenol) analytical standard](/deepweb/assets/sigmaaldrich/product/structures/304/181/a4c56697-e329-4ac0-b480-04de3e35f593/640/a4c56697-e329-4ac0-b480-04de3e35f593.png)
2,2′-Methylenebis(6-tert-butyl-4-methylphenol)
![2,2′-Methylenebis(6-tert-butyl-4-methylphenol) analytical standard](/deepweb/assets/sigmaaldrich/product/structures/304/181/a4c56697-e329-4ac0-b480-04de3e35f593/640/a4c56697-e329-4ac0-b480-04de3e35f593.png)
Linear Formula:
CH2[C6H2[C(CH3)3](CH3)OH]2
CAS Number:
Molecular Weight:
340.50
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing