![1-Ethyl-3-methylimidazolium hexafluorophosphate ≥99%, <500 ppm H2O](/deepweb/assets/sigmaaldrich/product/structures/100/575/2e598925-7548-4d58-ab50-ad8610f2b1fc/640/2e598925-7548-4d58-ab50-ad8610f2b1fc.png)
1-Ethyl-3-methylimidazolium hexafluorophosphate
![1-Ethyl-3-methylimidazolium hexafluorophosphate ≥99%, <500 ppm H2O](/deepweb/assets/sigmaaldrich/product/structures/100/575/2e598925-7548-4d58-ab50-ad8610f2b1fc/640/2e598925-7548-4d58-ab50-ad8610f2b1fc.png)
Synonym(s):
EMIMPF6
Empirical Formula (Hill Notation):
C6H11F6N2P
CAS Number:
Molecular Weight:
256.13
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Application
Showing 1-2 of 2
Compare
Product Number
Description
Pricing