Skip to Content
Merck
vibrant-m

17α-Dihydroequilin

vibrant-m
Synonym(s):
α-Equilol, Estra-1,3,5(10),7-tetraene-3,17α-diol
Empirical Formula (Hill Notation):
C18H22O2
CAS Number:
Molecular Weight:
270.37

Products

Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade