Skip to Content
Merck
1,2-O-Isopropylidene-α-D-xylofuranose 99%

1,2-O-Isopropylidene-α-D-xylofuranose

1,2-O-Isopropylidene-α-D-xylofuranose 99%
Empirical Formula (Hill Notation):
C8H14O5
CAS Number:
Molecular Weight:
190.19

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing