Product Name
Hydroquinine hydrobromide hydrate 98%
Product Number
341320
Product Brand
aldrich
CAS Number
207386-86-5
Molecular Weight
407.34 (anhydrous basis)
Test
Specification
APPEARANCE (COLOR)White
APPEARANCE (FORM)Powder
ARGENTOMETR. TITRATION97.5 - 102.5 %
PURITY (HPLC AREA %)≥ 97.5 %
OPTICAL ROTATION-100.0 ± 3.0 Degrees
CONCENTRATIONC=1.9, H2O
WATER≥ 2.1 %
INFRARED SPECTRUMCONFORMS TO STRUCTURE