Product Name
4-(Benzyloxy)benzaldehyde 97%
Product Number
123714
Product Brand
aldrich
CAS Number
4397-53-9
Molecular Weight
212.24
Test | Specification |
---|---|
APPEARANCE (COLOR) | White to Faint Yellow to Faint Red |
APPEARANCE (FORM) | Powder or Crystals |
PURITY (GC AREA %) | ≥ 96.5 % |
INFRARED SPECTRUM | CONFORMS TO STRUCTURE |