Skip to Content
Merck
All Photos(1)

Documents

T8018

Sigma-Aldrich

Thymidine 3′-monophosphate ammonium salt hydrate

~99% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H14N2O8P · xNH3 · yH2O
CAS Number:
Molecular Weight:
321.20 (anhydrous free acid basis)
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:

Assay

~99% (HPLC)

storage temp.

−20°C

SMILES string

N.O.CC1=CN([C@H]2C[C@H](OP(O)(O)=O)[C@@H](CO)O2)C(=O)NC1=O

InChI

1S/C10H15N2O8P.H3N.H2O/c1-5-3-12(10(15)11-9(5)14)8-2-6(7(4-13)19-8)20-21(16,17)18;;/h3,6-8,13H,2,4H2,1H3,(H,11,14,15)(H2,16,17,18);1H3;1H2/t6-,7+,8+;;/m0../s1

InChI key

AWIYUQSEPILHQT-ZJWYQBPBSA-N

Preparation Note

Enzymatically prepared.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D Bazile et al.
Biochemical and biophysical research communications, 159(2), 633-639 (1989-03-15)
DC-3F cells were submitted to electric square wave pulses in the presence of a d[alpha]octothymidylate 32P labelled in the 5' position. Radioactivity was incorporated in a voltage-dependent manner and reached a maximum for a field intensity of 1300-1400 V.cm-1. Growth

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service