Skip to Content
Merck
All Photos(1)

Key Documents

SAB5702227

Sigma-Aldrich

Anti-ABL2 Antibody, clone 2L9C6, Rabbit Monoclonal

Synonym(s):

ABLL, ARG

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
HUF 155,000.00

HUF 155,000.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 μL
HUF 155,000.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

HUF 155,000.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

material

colorless

clone

2L9C6, monoclonal

form

liquid

mol wt

128 kDa

species reactivity

rat, human, rat

concentration

0.47 mg/mL

technique(s)

western blot: 1:500 - 1:2000

color

colorless

isotype

IgG

immunogen sequence

KGYRMEQPEGCPPKVYELMRACWKWSPADRPSFAETHQAFETMFHDSSISEEVAEELGRAASSSSVVPYLPRLPILPSKTRTLKKQVENKENIEGAQDATE

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ABL2(27)

General description

This gene encodes a member of the Abelson family of nonreceptor tyrosine protein kinases. The protein is highly similar to the c-abl oncogene 1 protein, including the tyrosine kinase, SH2 and SH3 domains, and it plays a role in cytoskeletal rearrangements through its C-terminal F-actin- and microtubule-binding sequences. This gene is expressed in both normal and tumor cells, and is involved in translocation with the ets variant 6 gene in leukemia. Multiple alternatively spliced transcript variants encoding different protein isoforms have been found for this gene. [provided by RefSeq, Nov 2009]

Immunogen

A synthetic peptide corresponding to a sequence within amino acids 500-600 of human ABL2 (P42684).
PBS with 0.02% sodium azide,0.05% BSA,50% glycerol,pH7.3.

Application

WB

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service