Skip to Content
Merck
All Photos(1)

Documents

297577

Sigma-Aldrich

Inulin, 14C-carboxylated

solid

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352200

form

solid

mol wt

~5,000

extent of labeling

1-3 mCi per g

shipped in

dry ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Packaging

screw cap bottle

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service