Skip to Content
Merck
All Photos(1)

Key Documents

NIST333A

Molybdenum sulfide concentrate

NIST® SRM® 333a

Synonym(s):

Molybdenum(IV) sulfide, Molybdenum disulfide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
MoS2
CAS Number:
Molecular Weight:
160.07
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

Quality Level

form

powder

packaging

pkg of 1 pouch x 60 g

manufacturer/tradename

NIST®

density

5.06 g/mL at 25 °C (lit.)

application(s)

pharmaceutical (small molecule)

SMILES string

S=[Mo]=S

InChI

1S/Mo.2S

InChI key

CWQXQMHSOZUFJS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This Standard Reference Material (SRM) is a molybdenum sulfide (MoS2) concentrate from a commercial mining and refining process. SRM 333a is intended for use in the evaluation of chemical and instrumental methods of analysis. A unit of SRM 333a consists of one pouch containing approximately 60 g of powder. For more information, please refer to the COA and SDS.

SRM 333A_cert

SRM 333A_SDS

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service