Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

81799

Sigma-Aldrich

Propylphosphonic anhydride solution

technical, ~50% in ethyl acetate

Synonym(s):

1-Propanephosphonic anhydride solution, 2,4,6-Tripropyl-1,3,5,2,4,6-trioxatriphosphorinane-2,4,6-trioxide solution, PPACA, T3P®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H21O6P3
CAS Number:
Molecular Weight:
318.18
Beilstein:
5079255
MDL number:
UNSPSC Code:
12352100

Pricing and availability is not currently available.

grade

technical

concentration

~50% in ethyl acetate

SMILES string

CCCP1(=O)OP(=O)(CCC)OP(=O)(CCC)O1

InChI

1S/C9H21O6P3/c1-4-7-16(10)13-17(11,8-5-2)15-18(12,14-16)9-6-3/h4-9H2,1-3H3

InChI key

PAQZWJGSJMLPMG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

T3P is a registered trademark of Archimica GmbH

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service