Skip to Content
Merck
All Photos(1)

Key Documents

8.44007

Sigma-Aldrich

1,2-Butylene carbonate

for synthesis

Synonym(s):

1,2-Butylene carbonate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
116.12
EC Index Number:
403-780-4
Pricing and availability is not currently available.

vapor pressure

0.07 hPa ( 20 °C)

Quality Level

potency

>5000 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rabbit)

pH

6-8 (10 g/L in H2O)

kinematic viscosity

3 cSt

mp

-45 °C

transition temp

flash point 121 °C

density

1.14 g/cm3 at 25 °C

InChI

1S/C5H8O3/c1-2-4-3-7-5(6)8-4/h4H,2-3H2,1H3

InChI key

ZZXUZKXVROWEIF-UHFFFAOYSA-N

General description

for synthesis

Application

1,2-Butylene carbonate for synthesis. CAS 4437-85-8, pH 6 - 8 (10 g/l, H₂O).

Analysis Note

Evaporation Number: <0.01

Storage Class Code

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service