Skip to Content
Merck
All Photos(1)

Key Documents

ENAH942F3017

Enamine

ethyl 3-{[(tert-butoxy)carbonyl]amino}piperidine-4-carboxylate

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
HUF 165,794.00
100 MG
HUF 238,412.00
250 MG
HUF 333,260.00
500 MG
HUF 515,052.00
1 G
HUF 655,348.00

Vendor SKU: EN300-12715625-50MG

HUF 165,794.00


Ships in 14 days - from Aldrich Partner.


Select a Size

Change View
50 MG
HUF 165,794.00
100 MG
HUF 238,412.00
250 MG
HUF 333,260.00
500 MG
HUF 515,052.00
1 G
HUF 655,348.00

About This Item

CAS Number:
Molecular Weight:
272.34
MDL number:

Vendor SKU: EN300-12715625-50MG

HUF 165,794.00


Ships in 14 days - from Aldrich Partner.

Iupac Name

ethyl 3-((tert-butoxycarbonyl)amino)piperidine-4-carboxylate

Inchi Code

1S/C13H24N2O4/c1-5-18-11(16)9-6-7-14-8-10(9)15-12(17)19-13(2,3)4/h9-10,14H,5-8H2,1-4H3,(H,15,17)

InChI key

ICLFKLQIHHBWQG-UHFFFAOYSA-N

Purity

95

Country of Origin

UA

Product Link

https://www.enaminestore.com/catalog/EN300-12715625

MSDS

https://www.enaminestore.com/catalog/EN300-12715625


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service