Skip to Content
Merck
All Photos(1)

Key Documents

ENA497679429

Enamine

2-{2-oxo-1-oxa-4-thiaspiro[4.4]nonan-3-yl}acetic acid

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
HUF 88,236.00
250 MG
HUF 118,864.00
500 MG
HUF 208,278.00
1 G
HUF 265,582.00
2.5 G
HUF 503,196.00
5 G
HUF 736,858.00
10 G
HUF 1,084,634.00

Vendor SKU: EN300-261613-100MG

HUF 88,236.00


Ships in 14 days - from Aldrich Partner.


Select a Size

Change View
100 MG
HUF 88,236.00
250 MG
HUF 118,864.00
500 MG
HUF 208,278.00
1 G
HUF 265,582.00
2.5 G
HUF 503,196.00
5 G
HUF 736,858.00
10 G
HUF 1,084,634.00

About This Item

CAS Number:
Molecular Weight:
216.26
MDL number:

Vendor SKU: EN300-261613-100MG

HUF 88,236.00


Ships in 14 days - from Aldrich Partner.

mp

93-94

Iupac Name

2-(2-oxo-1-oxa-4-thiaspiro[4.4]nonan-3-yl)acetic acid

Inchi Code

1S/C9H12O4S/c10-7(11)5-6-8(12)13-9(14-6)3-1-2-4-9/h6H,1-5H2,(H,10,11)

InChI key

KZPNJNAEUVMDMA-UHFFFAOYSA-N

storage temp.

RT

Purity

95

Country of Origin

UA

Shipping Temperature

Normal

Physical Form

powder

Product Link

https://www.enaminestore.com/catalog/EN300-261613

Melting Point

93-94

MSDS

https://www.enaminestore.com/catalog/EN300-261613


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service