Skip to Content
Merck
All Photos(1)

Key Documents

ENA065085958

Enamine

(4-chlorophenyl)(1-methyl-1H-imidazol-2-yl)methanamine

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
HUF 58,102.00
250 MG
HUF 75,392.00
500 MG
HUF 108,984.00
1 G
HUF 135,166.00
2.5 G
HUF 223,592.00
5 G
HUF 370,310.00
10 G
HUF 539,752.00

Vendor SKU: EN300-14784-100MG

HUF 58,102.00


Ships in 14 days - from Aldrich Partner.


Select a Size

Change View
100 MG
HUF 58,102.00
250 MG
HUF 75,392.00
500 MG
HUF 108,984.00
1 G
HUF 135,166.00
2.5 G
HUF 223,592.00
5 G
HUF 370,310.00
10 G
HUF 539,752.00

About This Item

CAS Number:
Molecular Weight:
221.69
MDL number:

Vendor SKU: EN300-14784-100MG

HUF 58,102.00


Ships in 14 days - from Aldrich Partner.

Iupac Name

(4-chlorophenyl)(1-methyl-1H-imidazol-2-yl)methanamine

Inchi Code

1S/C11H12ClN3/c1-15-7-6-14-11(15)10(13)8-2-4-9(12)5-3-8/h2-7,10H,13H2,1H3

InChI key

OVCRVBWYZCVEHH-UHFFFAOYSA-N

storage temp.

4

Purity

95

Country of Origin

UA

Shipping Temperature

Normal

Physical Form

powder

Product Link

https://www.enaminestore.com/catalog/EN300-14784

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

MSDS

https://www.enaminestore.com/catalog/EN300-14784


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service