Skip to Content
Merck
All Photos(1)

Key Documents

ATEH984B2FC4

AstaTech, Inc.

5-CHLORONAPHTHALEN-1-OL

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
178.62
MDL number:

Iupac Name

5-chloro-1-naphthol

Inchi Code

1S/C10H7ClO/c11-9-5-1-4-8-7(9)3-2-6-10(8)12/h1-6,12H

InChI key

IPZJPYHEVWLBNH-UHFFFAOYSA-N

Purity

95%

Physical Form

Yellow to Brown Solid

Product Link

http://astatechinc.com/CPSResult.php?CRNO=241460

MSDS

http://astatechinc.com/Asta_MSDS.php?Catalog=E80257


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service