ATE458962906
AstaTech, Inc.
1-(PYRIDIN-2-YLCARBONYL)PIPERAZINE
About This Item
Iupac Name
1-(2-pyridinylcarbonyl)piperazine
Inchi Code
1S/C10H13N3O/c14-10(9-3-1-2-4-12-9)13-7-5-11-6-8-13/h1-4,11H,5-8H2
InChI key
IULDWWUGMGLWOB-UHFFFAOYSA-N
storage temp.
Purity
Shipping Temperature
Physical Form
Product Link
Signal Word
Warning
Hazard Statements
Precautionary statements
MSDS
http://astatechinc.com/Asta_MSDS.php?Catalog=20804
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service