ATE111598072
AstaTech, Inc.
2-METHOXY-4-PIPERAZIN-1-YL-PHENOL
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
2-methoxy-4-(1-piperazinyl)phenol
Inchi Code
1S/C11H16N2O2/c1-15-11-8-9(2-3-10(11)14)13-6-4-12-5-7-13/h2-3,8,12,14H,4-7H2,1H3
InChI key
JNYQZYVLEBQJSV-UHFFFAOYSA-N
storage temp.
2-8 C
Purity
97%
Country of Origin
CN
Shipping Temperature
Normal
Physical Form
Yellow to Brown Solid
Product Link
http://astatechinc.com/CPSResult.php?CRNO=7343
Signal Word
Warning
Hazard Statements
Precautionary statements
MSDS
http://astatechinc.com/Asta_MSDS.php?Catalog=63936
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service