Skip to Content
Merck
All Photos(1)

Key Documents

AMBH996F244E

Ambeed, Inc.

3-Methoxy-3-methylbut-1-yne

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
98.14
MDL number:

Iupac Name

1,1-dimethyl-2-propynyl methyl ether

Inchi Code

1S/C6H10O/c1-5-6(2,3)7-4/h1H,2-4H3

InChI key

ZYDXBIXQPLRRIK-UHFFFAOYSA-N

storage temp.

Sealed in dry,2-8C

Purity

95%

Country of Origin

CN

Shipping Temperature

Cold-chain transportation

Physical Form

liquid

Product Link

https://www.ambeed.com/products/13994-57-5.html

Synonym

3-Methoxy-3-methylbut-1-yne

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service