Skip to Content
Merck
All Photos(1)

Documents

W257109

Sigma-Aldrich

Hexyl 2-furoate

≥97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H16O3
CAS Number:
Molecular Weight:
196.24
FEMA Number:
2571
EC Number:
Council of Europe no.:
361
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
13.005

biological source

synthetic

grade

Kosher

Assay

≥97%

refractive index

n20/D 1.472 (lit.)

bp

105 °C/1 mmHg (lit.)

density

1.017 g/mL at 25 °C (lit.)

SMILES string

CCCCCCOC(=O)c1ccco1

InChI

1S/C11H16O3/c1-2-3-4-5-8-14-11(12)10-7-6-9-13-10/h6-7,9H,2-5,8H2,1H3

InChI key

KMLFVAGUWNPADU-UHFFFAOYSA-N

Other Notes

Download our Flavors and Fragrances Catalog to view our entire product line.

Subscribe to our Newsletter to keep up to date on our latest Flavors and Fragrances offerings.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service