Skip to Content
Merck
All Photos(1)

Documents

594652

Sigma-Aldrich

3-Methoxycarbonylphenylboronic acid pinacol ester

97%

Synonym(s):

3-Carbomethoxyphenylboronic acid pinacol ester, Methyl 3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H19BO4
CAS Number:
Molecular Weight:
262.11
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

91-95 °C (lit.)

SMILES string

COC(=O)c1cccc(c1)B2OC(C)(C)C(C)(C)O2

InChI

1S/C14H19BO4/c1-13(2)14(3,4)19-15(18-13)11-8-6-7-10(9-11)12(16)17-5/h6-9H,1-5H3

InChI key

JBJGSVBGUBATNH-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service