Skip to Content
Merck
All Photos(1)

Key Documents

527335

Sigma-Aldrich

PSS-Cyanopropyldimethylsilyloxy-Heptacyclopentyl substituted

Synonym(s):

(Cyanopropyldimethylsilyloxy)heptacyclopentylpentacyclooctasiloxane, Cyanopropyldimethylsilyloxy-POSS®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C41H75NO13Si9
CAS Number:
Molecular Weight:
1042.80
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

form

solid

mp

290 °C (dec.) (lit.)

SMILES string

C[Si](C)(CCCC#N)O[Si]12O[Si]3(O[Si](O1)(O[Si]45O[Si](O3)(O[Si](O4)(O[Si](O5)(C6CCCC6)C7CCCC7)C8CCCC8)C9CCCC9)C%10CCCC%10)O[Si](O2)(C%11CCCC%11)C%12CCCC%12

InChI

1S/C41H75NO13Si9/c1-56(2,34-18-17-33-42)43-62-46-58(37-23-7-8-24-37,38-25-9-10-26-38)47-64(55-62)51-60(40-29-13-14-30-40)48-59(39-27-11-12-28-39)44-57(35-19-3-4-20-35,36-21-5-6-22-36)45-63(49-59,50-60)53-61(52-62,54-64)41-31-15-16-32-41/h35-41H,3-32,34H2,1-2H3

InChI key

MBLNWXMDCNMJIP-UHFFFAOYSA-N

Legal Information

POSS is a registered trademark of Hybrid Plastics, Inc.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service