Merck
HomeProfessor Product PortalHeller Group – Professor Product Portal
Heller Group – Professor Product Portal
Professor Stephen Heller
Professor Stephen Heller
Professor Heller and coworkers are engaged in the development of mild and chemoselective acylation reactions using carbonylazole-derived reagents. To that end, they have developed a suite of carbonylimidazole derivatives for facile and chemoselective esterification (MImC, etc.) and amidation (WImC) of carboxylic acids, in collaboration with Professor Richmond Sarpong. These transformations are greatly accelerated by mildly acidic pyridinium salts. In combination with catalytic quantities of DBU, these reagents also effect selective N-acylation of indoles and oxazolidinones in the presence of more reactive functional groups such as anilines, amides, and phenolates.
Heller Group Website
Recent papers from the Heller Group
1.
Heller ST, Schultz EE, Sarpong R. 2012. Chemoselective N-Acylation of Indoles and Oxazolidinones with Carbonylazoles. Angew. Chem. Int. Ed.. 51(33):8304-8308. https://doi.org/10.1002/anie.201203976
2.
Heller ST, Fu T, Sarpong R. 2012. Dual Brønsted Acid/Nucleophilic Activation of Carbonylimidazole Derivatives. Org. Lett.. 14(8):1970-1973. https://doi.org/10.1021/ol300339q
3.
Heller ST, Sarpong R. 2010. Chemoselective Esterification and Amidation of Carboxylic Acids with Imidazole Carbamates and Ureas. Org. Lett.. 12(20):4572-4575. https://doi.org/10.1021/ol1018882
Loading
DISCIPLINES
  • Organic Synthetic Chemistry
  • Catalysis
Professor Product Portal Index