Merck
HomeProfessor Product PortalBuchwald Group – Professor Product Portal
Buchwald Group – Professor Product Portal
Stephen Buchwald
Professor Stephen Buchwald
The Buchwald group has developed a series of highly active and versatile palladium precatalysts and biarylphosphine ligands used in cross-coupling reactions for the formation of C-C, C–N, C–O, C–F, C–CF3, and C–S bonds. The ligands are electron-rich, and highly tunable to provide catalyst systems with a diverse scope, high stability and reactivity. Furthermore, the new series of precatalysts are air-, moisture and thermally-stable and display good solubility in common organic solvents. The use of precatalysts ensures the efficient generation of the active catalytic species and allows one to accurately adjust the ligand:palladium ratio. The ligands, precatalysts and methodology developed in the Buchwald group are user friendly and have rendered previously difficult cross couplings reactions, much easier to achieve.
Buchwald Group Website
Recent papers from the Buchwald Group
1.
Lee HG, Milner PJ, Buchwald SL. 2014. Pd-Catalyzed Nucleophilic Fluorination of Aryl Bromides. J. Am. Chem. Soc.. 136(10):3792-3795. https://doi.org/10.1021/ja5009739
2.
Su M, Hoshiya N, Buchwald SL. 2014. Palladium-Catalyzed Amination of Unprotected Five-Membered Heterocyclic Bromides. Org. Lett.. 16(3):832-835. https://doi.org/10.1021/ol4035947
3.
Surry DS, Buchwald SL. Dialkylbiaryl phosphines in Pd-catalyzed amination: a user's guide. Chem. Sci.. 2(1):27-50. https://doi.org/10.1039/c0sc00331j
Products available from the Buchwald Group