Merck
  • Cleavage of p21Cip1/Waf1 and p27Kip1 mediates apoptosis in endothelial cells through activation of Cdk2: role of a caspase cascade.
Cleavage of p21Cip1/Waf1 and p27Kip1 mediates apoptosis in endothelial cells through activation of Cdk2: role of a caspase cascade.
Molecular cell (1998-07-14)
B Levkau, H Koyama, E W Raines, B E Clurman, B Herren, K Orth, J M Roberts, R Ross
ABSTRACT
Apoptosis of human endothelial cells after growth factor deprivation is associated with rapid and dramatic up-regulation of cyclin A-associated cyclin-dependent kinase 2(cdk2) activity. In apoptotic cells, the C termini of the cdk inhibitors p21Cip1/Waf1 and p27Kip1 are truncated by specific cleavage. The enzyme involved in this cleavage is CPP32 and/or a CPP32-like caspase. After cleavage, p21Cip1/Waf1 loses its nuclear localization sequence and exits the nucleus. Cleavage of p21Cip1/Waf1 and p27Kip1 results in a substantial reduction in their association with nuclear cyclin-cdk2 complexes, leading to a dramatic induction of cdk2 activity. Dominant-negative cdk2, as well as a mutant of p21Cip1/Waf1 resistant to caspase cleavage, partially suppress apoptosis. These data suggest that cdk2 activation, through caspase-mediated cleavage of cdk inhibitors, may be instrumental in the execution of apoptosis following caspase activation.
Sigma-Aldrich
CDK2/Cyclin A2, active, GST tagged human, PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution
Sigma-Aldrich
CDK2/CyclinA1, active, GST tagged human, PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution