Merck
  • Identification and characterization of an endoplasmic reticulum localization motif.
Identification and characterization of an endoplasmic reticulum localization motif.
Biochemical and biophysical research communications (2016-03-24)
Yong-Wei Nie, Meng Wang, Ping Zhang, Lei Huan, Dong-Jun Liu
ABSTRACT
Sorting motifs are involved in the transport of diverse proteins. In the present study, we identified a hydrophobic peptide (WRPWRNFWWSIRVPWRRN) that was able to target enhanced green fluorescent protein- or DsRed2-enriched vesicular-like sub-compartments of the endoplasmic reticulum (ER). Analysis of mutation constructs revealed that the sequence WRPWRNFWW was responsible for the ER-targeting activity, and the arginine residue of the peptide is a critical determinant of ER localization. Results from co-immunoprecipitation, glutathione S-transferase pull-down, liquid chromatography-tandem mass spectrometry, and western blotting analyses demonstrated that this motif could bind with the γ2-COP subcomplex of coat protein complex I (COPI), which is involved in the retrieval and transport of ER-resident proteins from the Golgi apparatus to the ER. Overall, we report a new hydrophobic peptide that possesses an arginine-based ER localization motif, which can help elucidate the mechanisms of ER sorting mediated by COPI.
Product Description
Millipore
Glutathione High Capacity Magnetic Agarose Beads, (1:1 suspension in a 30% ethanol solution)