Merck
Clinical electrophysiologic effects of flecainide acetate.
Cardiovascular drugs and therapy (1988-03-01)
D Mehta, A J Camm, D E Ward
ABSTRACT
Flecainide acetate depresses the rate of depolarization of action potential (Vmax), the so-called "membrane stabilizing action." In the intact heart it has a unique profile of substantial effect on conduction with modest effect on refractoriness. After intravenous administration, clinical electrophysiologic studies show that conduction through atrial myocardium, atrioventricular (AV) node, His-Purkinje system, and ventricular myocardium is depressed, the most prominent effect being on the His-Purkinje system. Refractorines of the normal atrial and AV nodal myocardium is not prolonged while that of the ventricular muscle is slightly increased. Atrial fibrillation (60% to 70%), atrial tachycardia (90% to 100%), and nodal and AV tachycardia (80% to 90%) are generally terminated, while flutter is usually slowed, but in a small proportion of patients (10% to 20%) might be terminated by the intravenous use of flecainide acetate. This drug has also been shown to be effective in terminating stable ventricular tachycardia (70%). However, it appears to be slightly less effective in suppressing inducibility of ventricular arrhythmias. Administered orally, flecainide is very effective in decreasing ventricular ectopic activity (80% to 95%) and nonsustained ventricular tachycardia. Thus, flecainide has a wide range of antiarrhythmic properties, making it a useful agent in the management of a variety of supraventricular and ventricular arrhythmias. In a small proportion of patients, however, its use can lead to apparent arrhythmogenic effects, the most dangerous being exacerbation of ventricular tachycardia.
Sigma-Aldrich
Flecainide acetate salt
Pricing and availability is not currently available.
Flecainide acetate, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.