Merck
  • Ionic liquid as an efficient promoting medium for fixation of CO2: clean synthesis of alpha-methylene cyclic carbonates from CO2 and propargyl alcohols catalyzed by metal salts under mild conditions.
Ionic liquid as an efficient promoting medium for fixation of CO2: clean synthesis of alpha-methylene cyclic carbonates from CO2 and propargyl alcohols catalyzed by metal salts under mild conditions.
The Journal of organic chemistry (2004-01-17)
Yanlong Gu, Feng Shi, Youquan Deng
ABSTRACT
Reactions of propargylic alcohols with CO(2) in a [BMIm][PhSO(3)]/CuCl catalytic system to produce the corresponding alpha-methylene cyclic carbonates were conducted with high yields. Mild reaction conditions, enhanced rates, improved yields, and recyclable ionic liquid catalyst systems are the remarkable features exhibited in this process. Furthermore, the use of large amounts of tertiary amines as well as nitrogen-containing organic solvent as employed in previously studies was avoided.
Sigma-Aldrich
2-Phenyl-3-butyn-2-ol, ≥98%
Sigma-Aldrich
3-Methyl-1-pentyn-3-ol, 98%
Sigma-Aldrich
3,5-Dimethyl-1-hexyn-3-ol, 98%