Merck
  • Highly stable and flexible silver nanowire-graphene hybrid transparent conducting electrodes for emerging optoelectronic devices.
Highly stable and flexible silver nanowire-graphene hybrid transparent conducting electrodes for emerging optoelectronic devices.
Nanoscale (2013-07-12)
Donghwa Lee, Hyungjin Lee, Yumi Ahn, Youngjun Jeong, Dae-Young Lee, Youngu Lee
ABSTRACT
A new AgNW-graphene hybrid transparent conducting electrode (TCE) was prepared by dry-transferring a chemical vapor deposition (CVD)-grown monolayer graphene onto a pristine AgNW TCE. The AgNW-graphene hybrid TCE exhibited excellent optical and electrical properties as well as mechanical flexibility. The AgNW-graphene hybrid TCE showed highly enhanced thermal oxidation and chemical stabilities because of the superior gas-barrier property of the graphene protection layer. Furthermore, the organic solar cells with the AgNW-graphene hybrid TCE showed excellent photovoltaic performance as well as superior long-term stability under ambient conditions.
Product Description
Supelco
Poly(methyl methacrylate), analytical standard, for GPC, 4,000
Supelco
Poly(methyl methacrylate), analytical standard, for GPC, 50,000
Supelco
Poly(methyl methacrylate), analytical standard, for GPC, 20,000
Supelco
Poly(methyl methacrylate), analytical standard, for GPC, 10,000
Sigma-Aldrich
Poly(methyl methacrylate)
Sigma-Aldrich
Poly(methyl methacrylate), average Mw ~350,000 by GPC
Sigma-Aldrich
Poly(methyl methacrylate)
Sigma-Aldrich
Poly(methyl methacrylate), average Mw ~15,000 by GPC, powder
Supelco
Poly(methyl methacrylate), analytical standard, for GPC, average Mw 97,000 (Typical), average Mn 46,000 (Typical)
Supelco
Poly(methyl methacrylate), analytical standard, for GPC, 2,000
Supelco
Poly(methyl methacrylate), analytical standard, for GPC, 100,000
Supelco
Poly(methyl methacrylate), analytical standard, for GPC, 8,000
Supelco
Poly(methyl methacrylate), analytical standard, for GPC, 2,480,000