Merck
  • Implication that potassium flux and increase in intracellular calcium are necessary for the initiation of sperm motility in salmonid fishes.
Implication that potassium flux and increase in intracellular calcium are necessary for the initiation of sperm motility in salmonid fishes.
Molecular reproduction and development (1994-12-01)
S Tanimoto, Y Kudo, T Nakazawa, M Morisawa
ABSTRACT
Flux of K+ and changes in intracellular Ca2+ in the sperm of salmonid fishes were measured with spectrophotometry, ion electrode, microscopic fluorometry, and radioisotope accumulation. Release of K+ occurred at the initiation of sperm motility which is induced by decrease in external K+ and the K+ efflux and sperm motility were inhibited by K+ channel blockers. Intracellular Ca2+ increased within a short period in K(+)-free condition, and the accumulation of 45Ca in sperm cells was higher in motile sperm than that in immotile sperm. The efflux of K+ and the increase in intracellular Ca2+ were suppressed when external K+ concentration increased, i.e., sperm remained immotile. These results suggest that efflux of K+ through K+ channel and subsequent increase in intracellular Ca2+ are prerequisite for the initiation of sperm motility.
Product Description
Supelco
Fentin hydroxide, PESTANAL®, analytical standard
Sigma-Aldrich
Triphenyltin hydroxide