Merck
  • Quantitative determination of lead in mixtures of lead(II) halides using solid-state 207Pb NMR spectroscopy.
Quantitative determination of lead in mixtures of lead(II) halides using solid-state 207Pb NMR spectroscopy.
The Analyst (2006-10-27)
Alicia Glatfelter, Nicole Stephenson, Shi Bai, Cecil Dybowski, Dale L Perry
ABSTRACT
We demonstrate a multi-spectrum technique for facile, quantitative determination of lead in solid materials using solid-state (207)Pb NMR that avoids the major problem of uniform excitation across a wide spectral range; the method can be employed without chemical separation or other chemical manipulations and without any prior sample preparation, resulting in a non-destructive analysis, and producing results that are in agreement with gravimetric analyses of mixed samples of the lead halides.
Sigma-Aldrich
Lead(II) fluoride, 99.99% trace metals basis
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPrice
1 kg
Please contact Customer Service for Availability
€80.10
To order products, please contact your local dealer.
Sigma-Aldrich
Lead(II) fluoride, powder, ≥99%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPrice
1 kg
Please contact Customer Service for Availability
€80.10
To order products, please contact your local dealer.