Merck
[The late effects of exposure to lead fluoride].
Gigiena i sanitariia (1992-03-01)
A S Filin, A I Fadeev, A A Silaev
ABSTRACT
Effect of lead fluoride dust on the micronuclear reticulocyte content in the bone marrow was shown. Some functional changes in the testicles and increase in the fetal death rate in exposure to 50 mg/m3 of lead fluoride dust were noted.
Sigma-Aldrich
Lead(II) fluoride, 99.99% trace metals basis
Sigma-Aldrich
Lead(II) fluoride, powder, ≥99%