Merck
  • Titania-supported silver-based bimetallic nanoparticles as photocatalysts.
Titania-supported silver-based bimetallic nanoparticles as photocatalysts.
Environmental science and pollution research international (2012-11-20)
M A Barakat, R I Al-Hutailah, M H Hashim, E Qayyum, J N Kuhn
ABSTRACT
Photocatalytic process has shown recently a great potential as an environmental friendly and clean remediation technology for organic pollutants in wastewater. This work described the synthesis of silver-based bimetallic nanoparticles using colloid chemistry and the subsequent immobilization onto titania to form composite photocatalytic materials (titania-supported Ag-Pt nanoparticles). The photocatalysts were characterized by X-ray diffraction, electron microscopy, and nitrogen physisorption. The catalytic activity of the photocatalysts was evaluated by photocatalytic degradation of phenol and 2-chlorophenol (2-CP) in synthetic wastewater solutions. The photocatalytic processes were conducted in a batch photoreactor containing appropriate solutions of phenol and 2-CP with UV irradiation of 450 W. UV-visible spectrophotometer was used for analyzing the concentration of phenol and 2-CP in solutions. Parameters affecting the photocatalytic process such as the solution pH, phenol and 2-CP concentrations, and catalyst concentration were investigated. The results obtained revealed that TiO(2)-supported Ag/Pt nanoparticles showed a higher activity for UV-photocatalytic degradation of both phenol and 2-CP pollutants in the solution (as compared to the plain rutile TiO(2)). The photodegradation processes were optimized by the 0.5-g/L catalyst with a pollutant concentration of 50 mg/L for all the samples. Complete degradation for both phenol and 2-CP was achieved after 120 min.
Sigma-Aldrich
2-Chlorophenol, 98%
Sigma-Aldrich
2-Chlorophenol, ≥99%
Supelco
2-Chlorophenol, PESTANAL®, analytical standard