Merck
pH-changes during intracoronal bleaching: an in vivo study.
The journal of contemporary dental practice (2011-12-22)
Daphne Camara Barcellos, Alessandra Buhler Borges, Regina Celia dos Santos Pinto Silva, Luciana Maria Ribeiro, Cesar Rogerio Pucci, Carlos Rocha Gomes Torres
ABSTRACT
This study aimed to measure pH changes during 14 days intracoronal bleaching with hydrogen peroxide/sodium perborate and carbamide peroxide/sodium perborate. Twenty patients presenting endodontically treated central maxillary incisors with color alterations were divided in two groups (n = 10): Group CP + SP: 37% carbamide peroxide + sodium perborate paste; Group HP + SP: 30% hydrogen peroxide + sodium perborate paste. The pH values were measured using a digital microprocessor at different times: Baseline, 2, 7 and 14 days. Data were analyzed with two-way ANOVA followed by Tukey's test (α = 0.05). ANOVA showed p < 0.00 which indicated significant difference between the groups. The mean values (± sd) and the results of the Tukey´s test were: HP + SP/14 days-7.98 (±0.58)a; HP + SP/7 days-8.59 (±0.18)b; HP + SP/2 days- 8.83 (±0.32)bc; HP + SP/Baseline-8.83 (±0.01)bc; CP + SP/ Baseline-8.89 (±0.01)bc; CP + SP/14 days-9.11 (±0.58)cd; CP + SP/7 days-9.54 (±0.16)de; CP + SP/2 days-9.66 (±0.08) de. The group HP + SP resulted in significantly lower pH values compared with group CP + SP. It can be concluded that both associations showed alkaline pH values; however, there was significant reduction in the pH values of the 30% hydrogen peroxide associated with sodium perborate after 14 days. The association of hydrogen peroxide and carbamide peroxide with sodium perborate paste presented alkaline characteristics during the 14-day evaluated period. Thus, regarding pH changes, both associations can be considered safe as intracoronal bleaching agents.
Sigma-Aldrich
Sodium perborate tetrahydrate, 9-11% available Oxygen
Sigma-Aldrich
Sodium perborate tetrahydrate, purum p.a., 96% (RT)
Sigma-Aldrich
Sodium perborate monohydrate, powder, 20-100 mesh