Merck
  • Effects of molecular structures on reduction properties of formyl groups in chlorophylls and pheophytins prepared from oxygenic photosynthetic organisms.
Effects of molecular structures on reduction properties of formyl groups in chlorophylls and pheophytins prepared from oxygenic photosynthetic organisms.
Bioorganic & medicinal chemistry (2011-06-15)
Kana Sadaoka, Shigenori Kashimura, Yoshitaka Saga
ABSTRACT
Reduction of the 7-formyl groups in chlorophyll (Chl) b and its demetalated compound pheophytin (Phe) b was kinetically analyzed by using tert-butylamine-borane complex (t-BuNH(2)·BH(3)), and was compared with that of the 3-formyl groups in Chl d and Phe d. Reduction kinetics of the 7-formyl group in Chl b was similar to that in Phe b in dichloromethane containing 5mM t-BuNH(2)·BH(3). Little difference of the reduction kinetics of the 7-formyl groups between Chl b and Phe b was in sharp contrast to the reduction kinetics of the 3-formyl groups in Chl d and Phe d: the 3-formyl group in Phe d was reduced 5.3-fold faster than that in Chl d. The 7-formyl groups in Chl b and Phe b were reduced more slowly than the 3-formyl groups in Chl d and Phe d, respectively. The difference of the reactivity between the 3- and 7-formyl groups was in line with (13)C NMR measurements of chlorophyllous pigments, in which the chemical shifts of carbon atoms in the 7-formyl groups of Chl b and Phe b were high-field shifted compared with those in the 3-formyl groups of Chl d and Phe d, respectively. These indicate that the 7-formyl groups in chlorophyllous pigments were less reactive for reduction to the corresponding hydroxymethyl groups than the 3-formyl groups due to the difference in electronic states of the formyl groups in the A- and B-rings of the chlorin macrocycle.
Sigma-Aldrich
tert-Butylamine, 98%
Sigma-Aldrich
tert-Butylamine, ≥99.5%
Sigma-Aldrich
tert-Butylamine hydrochloride, ≥98.0% (AT)