Merck
  • Regenerative periodontal surgery in interproximal intrabony defects with biodegradable barriers.
Regenerative periodontal surgery in interproximal intrabony defects with biodegradable barriers.
Journal of clinical periodontology (2000-04-01)
C E Dörfer, T S Kim, H Steinbrenner, R Holle, P Eickholz
ABSTRACT
The comparison of the effects of guided-tissue regeneration (GTR) using 2 different biodegradable barriers (polylactide acetyltributyl citrate; polydioxanon) in 3- and 2-wall intrabony defects. The polydioxanon barrier is an experimental membrane for GTR therapy that consists of an continuous occlusive barrier that has a layer of slings on the side that is meant to face the mucoperiosteal flap. 15 patients provided 15 pairs of similar contralateral periodontal defects: 12 predominantly 2-wall and 18 predominantly 3-wall intrabony defects. Each defect was randomly assigned to treatment with either polylactide acetyltributyl citrate (control [c]) or polydioxanon (test [t]) devices. At baseline and 6 months after surgery, clinical measurements (P1I, GI, PPD, PAL-V) were performed. Barrier exposure was commonly observed in both groups (control/test): 5/4 after 7 days, 9/11 after 14 days and 11/12 after 28 days postsurgically. 4 weeks after surgery, 77% of all barriers were exposed to some extent. However, both treatments revealed a significant GI reduction (p<0.05), PPD reduction [-4.63+/-1.85 mm (t), -4.17+/-1.89 mm (c); p<0.001] and PAL-V gain [3.97+/-1.17 mm (t), 3.40 mm+/-1.40 (c); p<0.001] 6 months after surgery. Regarding GI and PPD reduction as well as PAL-V gain, there were neither statistically significant nor clinically relevant differences between test and control: similar clinical results were found 6 months after surgical treatment using both biodegradable barriers. Based on the results of the present study, the use of both biodegradable barriers in GTR therapy may be recommended.
Product Description
Supelco
Tributyl 2-acetylcitrate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Tributyl 2-acetylcitrate, ≥98%, FG
Sigma-Aldrich
Tributyl O-acetylcitrate, 98%