Merck
  • Engineering the substrate specificity of a thermophilic penicillin acylase from thermus thermophilus.
Engineering the substrate specificity of a thermophilic penicillin acylase from thermus thermophilus.
Applied and environmental microbiology (2012-12-25)
Leticia L Torres, Angel Cantero, Mercedes del Valle, Anabel Marina, Fernando López-Gallego, José M Guisán, José Berenguer, Aurelio Hidalgo
ABSTRACT
A homologue of the Escherichia coli penicillin acylase is encoded in the genomes of several thermophiles, including in different Thermus thermophilus strains. Although the natural substrate of this enzyme is not known, this acylase shows a marked preference for penicillin K over penicillin G. Three-dimensional models were created in which the catalytic residues and the substrate binding pocket were identified. Through rational redesign, residues were replaced to mimic the aromatic binding site of the E. coli penicillin G acylase. A set of enzyme variants containing between one and four amino acid replacements was generated, with altered catalytic properties in the hydrolyses of penicillins K and G. The introduction of a single phenylalanine residue in position α188, α189, or β24 improved the K(m) for penicillin G between 9- and 12-fold, and the catalytic efficiency of these variants for penicillin G was improved up to 6.6-fold. Structural models, as well as docking analyses, can predict the positioning of penicillins G and K for catalysis and can demonstrate how binding in a productive pose is compromised when more than one bulky phenylalanine residue is introduced into the active site.
Sigma-Aldrich
Penicillin G sodium salt, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Penicillin G sodium salt, ~1650 U/mg
Sigma-Aldrich
Penicillin G sodium salt, 96.0-102.0%
Sigma-Aldrich
Penicillin Amidase from Escherichia coli, 5-10 units/mg protein
Supelco
Penicillin G potassium salt, VETRANAL®, analytical standard
Sigma-Aldrich
Penicillin G potassium salt, 95.0 - 102.0%
Sigma-Aldrich
Penicillin G potassium salt
Sigma-Aldrich
Penicillin G potassium salt, powder, BioReagent, suitable for cell culture