Merck
  • Assessment of Tissue Expression of the Oxytocin-Vasopressin Pathway in the Placenta of Women with a First-Episode Psychosis during Pregnancy.
Assessment of Tissue Expression of the Oxytocin-Vasopressin Pathway in the Placenta of Women with a First-Episode Psychosis during Pregnancy.
International journal of molecular sciences (2023-06-28)
Miguel A Ortega, Cielo García-Montero, Óscar Fraile-Martinez, Diego De Leon-Oliva, Diego Liviu Boaru, Coral Bravo, Juan A De Leon-Luis, Miguel A Saez, Angel Asúnsolo, Ignacio Romero-Gerechter, Alejandro Sanz-Giancola, Raul Diaz-Pedrero, Laura Lopez-Gonzalez, Luis G Guijarro, Silvestra Barrena-Blázquez, Julia Bujan, Natalio García-Honduvilla, Melchor Alvarez-Mon, Miguel Ángel Alvarez-Mon, Guillermo Lahera
ABSTRACT
Psychosis refers to a mental health condition characterized by a loss of touch with reality, comprising delusions, hallucinations, disorganized thought, disorganized behavior, catatonia, and negative symptoms. A first-episode psychosis (FEP) is a rare condition that can trigger adverse outcomes both for the mother and newborn. Previously, we demonstrated the existence of histopathological changes in the placenta of pregnant women who suffer an FEP in pregnancy. Altered levels of oxytocin (OXT) and vasopressin (AVP) have been detected in patients who manifested an FEP, whereas abnormal placental expression of these hormones and their receptors (OXTR and AVPR1A) has been proven in different obstetric complications. However, the precise role and expression of these components in the placenta of women after an FEP have not been studied yet. Thus, the purpose of the present study was to analyze the gene and protein expression, using RT-qPCR and immunohistochemistry (IHC), of OXT, OXTR, AVP, and AVPR1a in the placental tissue of pregnant women after an FEP in comparison to pregnant women without any health complication (HC-PW). Our results showed increased gene and protein expression of OXT, AVP, OXTR, and AVPR1A in the placental tissue of pregnant women who suffer an FEP. Therefore, our study suggests that an FEP during pregnancy may be associated with an abnormal paracrine/endocrine activity of the placenta, which can negatively affect the maternofetal wellbeing. Nevertheless, additional research is required to validate our findings and ascertain any potential implications of the observed alterations.
Sigma-Aldrich
Anti-Rabbit IgG (γ-chain specific)−Biotin antibody, Mouse monoclonal, clone RG-96, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Mouse IgG (whole molecule)–FITC antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Neurophysin 2/NP-AVP Antibody, clone PS 41, clone PS 41, from mouse