Merck
Expression of high-affinity human antibody fragments in bacteria.
Nature protocols (2012-02-04)
Romain Rouet, David Lowe, Kip Dudgeon, Brendan Roome, Peter Schofield, David Langley, John Andrews, Peter Whitfeld, Lutz Jermutus, Daniel Christ
ABSTRACT
Here we describe protocols for the expression of human antibody fragments in Escherichia coli. Antigen-specific clones are identified by soluble fragment ELISA and concentrated by periplasmic preparation. They are then further purified by affinity chromatography. This article provides an overview of expression and purification strategies for human antibody fragments, as well as detailed protocols for the identification of high-affinity binders and for affinity maturation.
Sigma-Aldrich
Trypsin from bovine pancreas, TPCK Treated, essentially salt-free, lyophilized powder, ≥10,000 BAEE units/mg protein
Sigma-Aldrich
ExtrAvidin®−Peroxidase, buffered aqueous solution
Sigma-Aldrich
Anti-c-Myc−Biotin antibody, Mouse monoclonal, clone 9E10, purified from hybridoma cell culture