Merck
  • Dietary Fructose Enhances the Ability of Low Concentrations of Angiotensin II to Stimulate Proximal Tubule Na⁺ Reabsorption.
Dietary Fructose Enhances the Ability of Low Concentrations of Angiotensin II to Stimulate Proximal Tubule Na⁺ Reabsorption.
Nutrients (2017-08-17)
Agustin Gonzalez-Vicente, Pablo D Cabral, Nancy J Hong, Jessica Asirwatham, Nianxin Yang, Jessica M Berthiaume, Fernando P Dominici, Jeffrey L Garvin
ABSTRACT
Fructose-enriched diets cause salt-sensitive hypertension. Proximal tubules (PTs) reabsorb 70% of the water and salt filtered through the glomerulus. Angiotensin II (Ang II) regulates this process. Normally, dietary salt reduces Ang II allowing the kidney to excrete more salt, thereby preventing hypertension. We hypothesized that fructose-enriched diets enhance the ability of low concentrations of Ang II to stimulate PT transport. We measured the effects of a low concentration of Ang II (10-12 mol/L) on transport-related oxygen consumption (QO₂), and Na/K-ATPase and Na/H-exchange (NHE) activities and expression in PTs from rats consuming tap water (Control) or 20% fructose (FRUC). In FRUC-treated PTs, Ang II increased QO₂ by 14.9 ± 1.3 nmol/mg/min (p < 0.01) but had no effect in Controls. FRUC elevated NHE3 expression by 19 ± 3% (p < 0.004) but not Na/K-ATPase expression. Ang II stimulated NHE activity in FRUC PT (Δ + 0.7 ± 0.1 Arbitrary Fluorescent units (AFU)/s, p < 0.01) but not in Controls. Na/K-ATPase activity was not affected. The PKC inhibitor Gö6976 blocked the ability of FRUC to augment the actions of Ang II. FRUC did not alter the inhibitory effect of dopamine on NHE activity. We conclude that dietary fructose increases the ability of low concentrations of Ang II to stimulate PT Na reabsorption via effects on NHE.
Sigma-Aldrich
Anti-Cytoplasmic FMR1-interacting protein 1 Antibody, from rabbit, purified by affinity chromatography
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution