Merck
  • The human GCOM1 complex gene interacts with the NMDA receptor and internexin-alpha.
The human GCOM1 complex gene interacts with the NMDA receptor and internexin-alpha.
Gene (2018-01-18)
Raymond S Roginski, Chi W Lau, Phillip P Santoiemma, Sara J Weaver, Peicheng Du, Patricia Soteropoulos, Jay Yang
ABSTRACT
The known functions of the human GCOM1 complex hub gene include transcription elongation and the intercalated disk of cardiac myocytes. However, in all likelihood, the gene's most interesting, and thus far least understood, roles will be found in the central nervous system. To investigate the functions of the GCOM1 gene in the CNS, we have cloned human and rat brain cDNAs encoding novel, 105 kDa GCOM1 combined (Gcom) proteins, designated Gcom15, and identified a new group of GCOM1 interacting genes, termed Gints, from yeast two-hybrid (Y2H) screens. We showed that Gcom15 interacts with the NR1 subunit of the NMDA receptor by co-expression in heterologous cells, in which we observed bi-directional co-immunoprecipitation of human Gcom15 and murine NR1. Our Y2H screens revealed 27 novel GCOM1 interacting genes, many of which are synaptic proteins and/or play roles in neurologic diseases. Finally, we showed, using rat brain protein preparations, that the Gint internexin-alpha (INA), a known interactor of the NMDAR, co-IPs with GCOM1 proteins, suggesting a GCOM1-GRIN1-INA interaction and a novel pathway that may be relevant to neuroprotection.
Sigma-Aldrich
Anti-Internexin α Antibody, serum, Chemicon®
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Anti-Synaptophysin Antibody, clone SP15, ascites fluid, clone SP15, Chemicon®