- Phe-Ala-based diazaspirocyclic lactam as nucleator of type II' β-turn.
Phe-Ala-based diazaspirocyclic lactam as nucleator of type II' β-turn.
The Journal of organic chemistry (2011-01-13)
Alessandro Sacchetti, Alessandra Silvani, Giordano Lesma, Tullio Pilati
PMID21222472
ABSTRACT
The synthesis of a novel Phe-Ala dipeptide mimic, built up on a diazaspirocyclic lactam core, is presented. This new scaffold was evaluated for conformational mimicry of reverse turn by combining molecular modeling, IR, NMR, and X-ray diffraction experiments. All these tools agree on the presence of a strong intramolecular hydrogen bond, thus demonstrating the ability of this spiro compound to act as a type II' β-turn inducer.
MATERIALS