
Verbascoside

Synonym(s):
Acetoside, Kusaginin, Verbascoside
Empirical Formula (Hill Notation):
C29H36O15
CAS Number:
Molecular Weight:
624.59
Products
Product Category
Brand
Feature
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Agency
Format
Research Area
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-5 of 5
Compare
Product Number
Description
Pricing